CELLTREAT Scientific Products 229697 Slippable Sterile Petri Dishes, 100 x 15 mm; 500/cs
Representative image only.
CELLTREAT Scientific Products 229697 Slippable Sterile Petri Dishes, 100 x 15 mm; 500/cs
CELLTREAT Scientific Products | Mfr: 229697| Item: 0193230
Optically clear for improved imaging and observation
₹33,400.00INR / Case of 500
₹33,400.00INR / Case of 500
Product Description
Includes
Lids.
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • Designed for automated filling and streaking systems
  • 15 to 16 mL working volume
More About this Item
These slippable dishes feature side arrows and ISO Market Target for automation. Flared lid skirt and squared corners provide easy one-handed operation, even while wearing gloves. Lids have molded spacers to reduce condensation and allow air circulation.
All dishes are supplied sterile (gamma irradiated) and come packaged 25 dishes/bag.
Sterile
Specifications
  • DescriptionSlippable Sterile Petri Dishes, 100 x 15 mm; 500/cs