CELLTREAT Scientific Products 229694 Slideable Sterile Petri Dishes, 100 x 15 mm; 500/cs
Representative image only.
CELLTREAT Scientific Products 229694 Slideable Sterile Petri Dishes, 100 x 15 mm; 500/cs
CELLTREAT Scientific Products | Mfr: 229694| Item: 0193228
Optically clear for improved imaging and observation
₹32,900.00INR / Case of 500
₹32,900.00INR / Case of 500
Product Description
Includes
Lids.
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • Specially designed to easily slide a single dish from a large stack
  • 15 to 16 mL working volume
More About this Item
A partial edge ring on base allows a single dish to be slid from a large stack while enhancing stability when stacked. Lids have molded spacers to reduce condensation and allow air circulation.
All dishes are supplied sterile (gamma irradiated) and come packaged 20 dishes/bag.
Sterile
Specifications
  • DescriptionSlideable Sterile Petri Dishes, 100 x 15 mm; 500/cs