CELLTREAT Scientific Products 229684 Quad Compartment Sterile Petri Dishes, 100 x 15 mm; 500/cs
Representative image only.
CELLTREAT Scientific Products 229684 Quad Compartment Sterile Petri Dishes, 100 x 15 mm; 500/cs
CELLTREAT Scientific Products | Mfr: 229684| Item: 0193224
Optically clear for improved imaging and observation
₹59,600.00INR / Case of 500
₹59,600.00INR / Case of 500
Product Description
Includes
Lids.
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • Run multiple experiments in one petri dish
  • 4 mL working volume/compartment
More About this Item
Use the compartment dishes to run simultaneous studies of organisms in a common environment. Lids have molded spacers to reduce condensation and allow air circulation. Stacking rings on the lids provide increased stability.
All dishes are supplied sterile (gamma irradiated) and come packaged 25 dishes/bag.
Sterile
Specifications
  • DescriptionQuad Compartment Sterile Petri Dishes, 100 x 15 mm; 500/cs