CELLTREAT Scientific Products 229663 Heavy-Duty Sterile Petri Dishes with Grip Ring, 60 x 15 mm; 500/cs
Representative image only.
CELLTREAT Scientific Products 229663 Heavy-Duty Sterile Petri Dishes with Grip Ring, 60 x 15 mm; 500/cs
CELLTREAT Scientific Products | Mfr: 229663| Item: 0193219
Optically clear for improved imaging and observation
₹37,200.00INR / Case of 500
₹37,200.00INR / Case of 500
Product Description
Includes
Lids.
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • 7 to 8 mL working volume.
More About this Item
These heavy-duty petri dishes are thicker and stronger for added durability. Lids have molded spacers to reduce condensation and allow air circulation. Stacking rings on the lids provide increased stability. A pronounced grip ring offers improved handling.
All dishes are supplied sterile (gamma irradiated) and come packaged 10 dishes/bag.
Sterile
Specifications
  • DescriptionHeavy-Duty Sterile Petri Dishes with Grip Ring, 60 x 15 mm; 500/cs