CELLTREAT Scientific Products 229660 Treated Sterile Petri Dishes with Grip Ring, 60 x 15 mm; 500/cs
Representative image only.
CELLTREAT Scientific Products 229660 Treated Sterile Petri Dishes with Grip Ring, 60 x 15 mm; 500/cs
CELLTREAT Scientific Products | Mfr: 229660| Item: 0193218
Optically clear for improved imaging and observation
₹68,500.00INR / Case of 500
₹68,500.00INR / Case of 500
Product Description
Includes
Lids.
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • 7 to 8 mL working volume
  • 21 cm² growth area
More About this Item
These treated petri dishes have a hydrophilic surface to promote cell attachment and growth. Lids contain molded spacers to allow gas exchange and stacking rings for increased stability. Bases feature a grip ring for improved handling.
All dishes are supplied sterile (gamma irradiated) and come packaged 10/re-sealable bag.
Sterile
Specifications
  • DescriptionTreated Sterile Petri Dishes with Grip Ring, 60 x 15 mm; 500/cs