CELLTREAT Scientific Products 229653 Heavy-Duty Sterile Petri Dishes, 150 x 20 mm; 100/cs
Representative image only.
CELLTREAT Scientific Products 229653 Heavy-Duty Sterile Petri Dishes, 150 x 20 mm; 100/cs
CELLTREAT Scientific Products | Mfr: 229653| Item: 0193216
Optically clear for improved imaging and observation
₹55,700.00INR / Case of 100
₹55,700.00INR / Case of 100
Product Description
Includes
Lids.
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • 25 to 28 mL working volume
More About this Item
These heavy-duty petri dishes are thicker and stronger for added durability. Lids have molded spacers to reduce condensation and allow air circulation. Stacking rings on the lids provide increased stability.
All dishes are supplied sterile (gamma irradiated) and come packaged 5 dishes/bag.
Sterile
Specifications
  • DescriptionHeavy-Duty Sterile Petri Dishes, 150 x 20 mm; 100/cs