CELLTREAT Scientific Products 229650 Treated Sterile Petri Dishes, 150 x 20 mm; 60/cs
Representative image only.
CELLTREAT Scientific Products 229650 Treated Sterile Petri Dishes, 150 x 20 mm; 60/cs
CELLTREAT Scientific Products | Mfr: 229650| Item: 0193214
Optically clear for improved imaging and observation
₹39,900.00INR / Case of 60
₹39,900.00INR / Case of 60
Product Description
Includes
Lids.
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • 25 to 28 mL working volume
  • 143 cm² growth area
More About this Item
These treated petri dishes have a hydrophilic surface to promote cell attachment and growth. Lids contain molded spacers to allow gas exchange and stacking rings for increased stability.
All dishes are supplied sterile (gamma irradiated) and come packaged 1/bag.
Sterile
Specifications
  • DescriptionTreated Sterile Petri Dishes, 150 x 20 mm; 60/cs