CELLTREAT Scientific Products 229638 Heavy-Duty Sterile Petri Dishes, 35 x 10 mm; 960/cs

Representative image only.

CELLTREAT Scientific Products 229638 Heavy-Duty Sterile Petri Dishes, 35 x 10 mm; 960/cs

CELLTREAT Scientific Products | Mfr: 229638| Item: 0193213

Optically clear for improved imaging and observation

₹52,900.00INR / Case of 960

₹52,900.00INR / Case of 960
Product Description

Includes

Lids.

Key Features

  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • Offset bottom for improved handling
  • 5 mL working volume

More About this Item

These heavy-duty petri dishes are thicker and stronger for added durability. Lids have molded spacers to reduce condensation and allow air circulation. Stacking rings on the lids provide increased stability.

All dishes are supplied sterile (gamma irradiated) and come packaged 10 dishes/bag.

Sterile
Specifications
  • DescriptionHeavy-Duty Sterile Petri Dishes, 35 x 10 mm; 960/cs
Reviews
Q&A