CELLTREAT Scientific Products 229620 Treated Sterile Petri Dishes with Grip Ring, 100 x 20 mm; 300/cs
Representative image only.
CELLTREAT Scientific Products 229620 Treated Sterile Petri Dishes with Grip Ring, 100 x 20 mm; 300/cs
CELLTREAT Scientific Products | Mfr: 229620| Item: 0193210
Optically clear for improved imaging and observation
₹70,200.00INR / Case of 300
₹70,200.00INR / Case of 300
Product Description
Includes
Lids.
Key Features
  • Consistent flatness allows for even media distribution
  • Manufactured from virgin polystyrene
  • All dishes are supplied sterile (gamma irradiated)
  • 15 to 16 mL working volume
  • 60.5 cm² growth area
More About this Item
These treated petri dishes have a hydrophilic surface to promote cell attachment and growth. Lids contain molded spacers to allow gas exchange and stacking rings for increased stability. A pronounced grip ring offers improved handling.
All dishes are supplied sterile (gamma irradiated) and come packaged 10 dishes/re-sealable bag.
Sterile
Specifications
  • DescriptionTreated Sterile Petri Dishes with Grip Ring, 100 x 20 mm; 300/cs