CELLTREAT Scientific Products 229582 Non-Treated Culture Roller Bottle, 1L, nonvented cap, sterile, 24/cs
Representative image only.
CELLTREAT Scientific Products 229582 Non-Treated Culture Roller Bottle, 1L, nonvented cap, sterile, 24/cs
CELLTREAT Scientific Products | Mfr: 229582| Item: 0183637
Uniform, hydrophilic surface promotes cell attachment
₹47,400.00INR / Case of 24
₹47,400.00INR / Case of 24
Buy It Together
Current Item
CELLTREAT Scientific Products 229582 Non-Treated Culture Roller Bottle, 1L, nonvented cap, sterile, 24/cs
CELLTREAT Scientific Products 229582 Non-Treated Culture Roller Bottle, 1L, nonvented cap, sterile, 24/cs
0183637
₹47,400.00INR / Case of 24
+ =
Total Price 60000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Uniform, hydrophilic surface for cell attachment
  • Nontreated roller bottles
  • Solid, nonvented caps
More About this Item
Bottles are made of clear polystyrene and feature easy-to-read black graduations. Each bottle is gamma irradiated and individually bagged. Caps have large knurls for easy opening.
Sterile
Specifications
  • MaterialPolystyrene
  • SterileYes
  • Capacity (mL)1000
  • DescriptionNon-Treated Culture Roller Bottle, 1L, nonvented cap, sterile, 24/cs