CELLTREAT Scientific Products 229572 96-Well Deep Well PP Storage Plates; 25/cs
Representative image only.
CELLTREAT Scientific Products 229572 96-Well Deep Well PP Storage Plates; 25/cs
CELLTREAT Scientific Products | Mfr: 229572| Item: 0195919
Great for growing bacterial cultures or storing compounds
₹58,100.00INR / Case of 25
₹58,100.00INR / Case of 25
Product Description
Key Features
  • Pyramid-bottom provide easy sample retrieval and improved mixing by creating a sloshing effect rather than a vortex when agitated
More About this Item
Polypropylene construction offers low binding surface to prevent samples from sticking to the side walls during elution and is chemically inert for combinatorial chemistry applications.
Specifications
  • Number of Wells96
  • Max Working Well Volume (mL)2
  • Well ShapePyramid-bottom
  • Description96-Well Deep Well PP Storage Plates; 25/cs