CELLTREAT Scientific Products 229387 Culture Roller Bottle, vented cap, sterile, 1900 sq. cm, 12/cs
Representative image only.
CELLTREAT Scientific Products 229387 Culture Roller Bottle, vented cap, sterile, 1900 sq. cm, 12/cs
CELLTREAT Scientific Products | Mfr: 229387| Item: 0183635
Uniform, hydrophilic surface promotes cell attachment
₹51,300.00INR / Case of 12
₹51,300.00INR / Case of 12
Buy It Together
Current Item
CELLTREAT Scientific Products 229387 Culture Roller Bottle, vented cap, sterile, 1900 sq. cm, 12/cs
CELLTREAT Scientific Products 229387 Culture Roller Bottle, vented cap, sterile, 1900 sq. cm, 12/cs
0183635
₹51,300.00INR / Case of 12
+ =
Total Price 63900.00 ADD 2 ITEMS TO CART
Product Description
Includes
Vented caps.
Key Features
  • Expanded surface roller bottles (ESRB) with ribbed surfaces increase growth area from 850 cm² to 1900 cm²
  • Vacuum gas plasma tissue culture treated
  • Vented caps—ideal for applications requiring continuous gas exchange
More About this Item
Bottles are made of clear polystyrene and feature easy-to-read black graduations. Each bottle is gamma irradiated and individually bagged. Caps have large knurls for easy opening.
Sterile
Specifications
  • MaterialPolystyrene
  • ClosureVented Cap
  • Culture Area (cm²)1900
  • SterileYes
  • Capacity (mL)2000
  • DescriptionCulture Roller Bottle, vented cap, sterile, 1900 sq. cm, 12/cs