CELLTREAT Scientific Products 229385 Culture Roller Bottle, vented cap, sterile, 850 sq. cm, 12/cs
Representative image only.
CELLTREAT Scientific Products 229385 Culture Roller Bottle, vented cap, sterile, 850 sq. cm, 12/cs
CELLTREAT Scientific Products | Mfr: 229385| Item: 0183633
Uniform, hydrophilic surface promotes cell attachment
₹30,500.00INR / Case of 12
₹30,500.00INR / Case of 12
Buy It Together
Current Item
CELLTREAT Scientific Products 229385 Culture Roller Bottle, vented cap, sterile, 850 sq. cm, 12/cs
CELLTREAT Scientific Products 229385 Culture Roller Bottle, vented cap, sterile, 850 sq. cm, 12/cs
0183633
₹30,500.00INR / Case of 12
+ =
Total Price 43100.00 ADD 2 ITEMS TO CART
Product Description
Includes
Vented caps
Key Features
  • Vacuum gas plasma tissue culture treated
  • Vented caps—ideal for applications requiring continuous gas exchange
More About this Item
Bottles are made of clear polystyrene and feature easy-to-read black graduations. Each bottle is gamma irradiated and individually bagged. Caps have large knurls for easy opening.
Sterile
Specifications
  • MaterialPolystyrene
  • ClosureVented Cap
  • Culture Area (cm²)850
  • SterileYes
  • Capacity (mL)2000
  • DescriptionCulture Roller Bottle, vented cap, sterile, 850 sq. cm, 12/cs