CELLTREAT Scientific Products 229384 Culture Roller Bottle, nonvented cap, sterile, 850 sq. cm,12/CS
Representative image only.
CELLTREAT Scientific Products 229384 Culture Roller Bottle, nonvented cap, sterile, 850 sq. cm,12/CS
CELLTREAT Scientific Products | Mfr: 229384| Item: 0183632
Uniform, hydrophilic surface promotes cell attachment
₹28,600.00INR / Case of 12
₹28,600.00INR / Case of 12
Buy It Together
Current Item
CELLTREAT Scientific Products 229384 Culture Roller Bottle, nonvented cap, sterile, 850 sq. cm,12/CS
CELLTREAT Scientific Products 229384 Culture Roller Bottle, nonvented cap, sterile, 850 sq. cm,12/CS
0183632
₹28,600.00INR / Case of 12
+ =
Total Price 41200.00 ADD 2 ITEMS TO CART
Product Description
Includes
Solid, nonvented caps.
Key Features
  • Vacuum gas plasma tissue culture treated
  • Solid, nonvented caps
More About this Item
Bottles are made of clear polystyrene and feature easy-to-read black graduations. Each bottle is gamma irradiated and individually bagged. Caps have large knurls for easy opening.
Sterile
Specifications
  • MaterialPolystyrene
  • ClosureNonvented Cap
  • Culture Area (cm²)850
  • SterileYes
  • Capacity (mL)2000
  • DescriptionCulture Roller Bottle, nonvented cap, sterile, 850 sq. cm, 12/cs