CELLTREAT Scientific Products 229382 Culture Roller Bottle, nonvented cap, sterile, 490 sq. cm, 24/cs
Representative image only.
CELLTREAT Scientific Products 229382 Culture Roller Bottle, nonvented cap, sterile, 490 sq. cm, 24/cs
CELLTREAT Scientific Products | Mfr: 229382| Item: 0183630
Uniform, hydrophilic surface promotes cell attachment
₹51,100.00INR / Case of 24
₹51,100.00INR / Case of 24
Buy It Together
Current Item
CELLTREAT Scientific Products 229382 Culture Roller Bottle, nonvented cap, sterile, 490 sq. cm, 24/cs
CELLTREAT Scientific Products 229382 Culture Roller Bottle, nonvented cap, sterile, 490 sq. cm, 24/cs
0183630
₹51,100.00INR / Case of 24
+ =
Total Price 63700.00 ADD 2 ITEMS TO CART
Product Description
Includes
Solid, nonvented caps.
Key Features
  • Vacuum gas plasma tissue culture treated
  • Solid, nonvented caps
More About this Item
Bottles are made of clear polystyrene and feature easy-to-read black graduations. Each bottle is gamma irradiated and individually bagged. Caps have large knurls for easy opening.
Sterile
Specifications
  • MaterialPolystyrene
  • Culture Area (cm²)490
  • SterileYes
  • Capacity (mL)1000
  • DescriptionCulture Roller Bottle, nonvented cap, sterile, 490 sq. cm, 24/cs