CE International Trading Mesh Screen, SS, 1.00 mm Sieve Opening, 16 mesh, 18" dia
Representative image only.
CE International Trading Mesh Screen, SS, 1.00 mm Sieve Opening, 16 mesh, 18" dia
CE International Trading | Mfr: LS450S005-#16MESH| Item: 5997949
Mesh screens separate oversized particles from your sample
₹59,300.00INR / Each
₹59,300.00INR / Each
Buy It Together
Current Item
CE International Trading Mesh Screen, SS, 1.00 mm Sieve Opening, 16 mesh, 18" dia
CE International Trading Mesh Screen, SS, 1.00 mm Sieve Opening, 16 mesh, 18" dia
5997949
₹59,300.00INR / Each
+ =
Total Price 94900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).
Specifications
  • MaterialStainless Steel
  • U.S. Standard Screen Size (No.)18
  • Tyler Standard Screen Size (mesh)16
  • Sieve Opening (mm)1
  • Outside Diameter (in)18
  • DescriptionMesh Screen, SS, 1.00 mm sieve opening, 16 mesh