CE International Trading Mesh Screen, Nylon, 0.425 mm Sieve Opening, 35 Mesh, 18" dia
Representative image only.
CE International Trading Mesh Screen, Nylon, 0.425 mm Sieve Opening, 35 Mesh, 18" dia
CE International Trading | Mfr: LS450N006-#35MESH| Item: 5997983
Mesh screens separate oversized particles from your sample
₹50,400.00INR / Each
₹50,400.00INR / Each
Buy It Together
Current Item
CE International Trading Mesh Screen, Nylon, 0.425 mm Sieve Opening, 35 Mesh, 18" dia
CE International Trading Mesh Screen, Nylon, 0.425 mm Sieve Opening, 35 Mesh, 18" dia
5997983
₹50,400.00INR / Each
+ =
Total Price 86000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).
Specifications
  • MaterialNylon
  • U.S. Standard Screen Size (No.)40
  • Tyler Standard Screen Size (mesh)35
  • Sieve Opening (mm)0.425
  • Outside Diameter (in)18
  • DescriptionMesh Screen, Nylon, 0.425 mm sieve opening, 35 mesh