CE International Trading Mesh Screen, Nylon, 0.045 mm Sieve Opening, 325 mesh, 18" dia
Representative image only.
CE International Trading Mesh Screen, Nylon, 0.045 mm Sieve Opening, 325 mesh, 18" dia
CE International Trading | Mfr: LS450N006-#325MESH| Item: 5997996
Mesh screens separate oversized particles from your sample
₹58,800.00INR / Each
₹58,800.00INR / Each
Buy It Together
Current Item
CE International Trading Mesh Screen, Nylon, 0.045 mm Sieve Opening, 325 mesh, 18" dia
CE International Trading Mesh Screen, Nylon, 0.045 mm Sieve Opening, 325 mesh, 18" dia
5997996
₹58,800.00INR / Each
+ =
Total Price 94400.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
More About this Item
Screens are 18" (45.7 cm) in diameter with holes for securing in the screen assembly (sold separately under part number 59998-54).
Specifications
  • MaterialNylon
  • U.S. Standard Screen Size (No.)325
  • Tyler Standard Screen Size (mesh)325
  • Sieve Opening (mm)0.044
  • Outside Diameter (in)18
  • DescriptionMesh Screen, Nylon, 0.045 mm sieve opening, 325 mesh