CE International Trading LS450N0028 Nylon Spiral Brush for Vibro-Separators, 60 mesh and finer
Representative image only.
CE International Trading LS450N0028 Nylon Spiral Brush for Vibro-Separators, 60 mesh and finer
CE International Trading | Mfr: LS450N0028| Item: 5997900
₹76,500.00INR / Each
₹76,500.00INR / Each
Product Description
Key Features
  • 1 month warranty
More About this Item
Nylon spiral brush cleans the screen and helps avoids clogs. Place brush on the screen cloth, and it automatically rotates due to the vibration. Note: Use with screen cloths finer than 60 mesh only.
1 Month Warranty
Specifications
  • DescriptionNylon Spiral Brush for Vibro-Separators, 60 mesh and finer
  • Warranty1 Month