Casella CEL-430.A1 Type I Sound Level Meter with Memory
Representative image only.
Casella CEL-430.A1 Type I Sound Level Meter with Memory
Casella | Mfr: CEL-620.A1| Item: 5050942
₹1,461,000.00INR / Each
for an additional
₹38,500.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹1,461,000.00INR / Each
Buy It Together
Current Item
Casella CEL-430.A1 Type I Sound Level Meter with Memory
Casella CEL-430.A1 Type I Sound Level Meter with Memory
5050942
₹1,461,000.00INR / Each
+
InnoCal NIST-Traceable Calibration; Sound Meter, Type II
1710153
₹38,500.00INR / Each
=
Total Price 1499500.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • FEATURES
    • Single 120 dB dynamic range covers all noise levels
    • Simple easy-to-use operation
    • Pre-configured setups for the most popular measurements
    • Available with ANSI/IEC type 1 and type 2 accuracy
    • Automatic recognition of calibrator when applied to microphone
    • Large custom LCD with automatic backlight
    • Memory for result storage
    • PC software program for archiving and printing results
    • Meter can be remotely configured for special measurements from
    • dB30 PC software
    • Fixed microphone and preamplifier for simplicity
  • 1 year warranty
More About this Item
The CEL430 Integrating sound level meter is a new design that encompasses many of the benefits of the existing CEL450 and CEL490 products in a simple and easy to use meter. All DSP technology ensures that the whole of the audio range of noise levels are covered by a single dynamic span. This eliminates the need to change ranges during a measurement and ensures that all the noise is captured faithfully and included in the overall result. All the most popular noise units are simultaneously calculated during the measurement and can be read off the large display or recorded for later download to the optional software package.
1 Year Warranty
Specifications
  • Meter ClassType 1
  • ComplianceANSI S1.4-1983: type 2; EN 651
  • DisplayLCD
  • DescriptionType I Sound Level Meter with Memory