Carver 2096CE Two-Gauge Manifold for Manual Hydraulic Press Test
Representative image only.
Carver 2096CE Two-Gauge Manifold for Manual Hydraulic Press Test
Carver | Mfr: 2096CE| Item: 5962061
₹292,000.00INR / Each
₹292,000.00INR / Each
Buy It Together
Current Item
Carver 2096CE Two-Gauge Manifold for Manual Hydraulic Press Test
Carver 2096CE Two-Gauge Manifold for Manual Hydraulic Press Test
5962061
₹292,000.00INR / Each
+
Carver 381002 Press Low Pressure Gauge; 0 to 2000 psi
5962062
₹212,000.00INR / Each
=
Total Price 504000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
Two-Gauge Manifold provides both low- and high-pressure gauge readings for greater precision. Hand-operated, two-gauge manifold includes a shut-off valve for the low-pressure gauge to protect it during high-pressure applications. Unit includes a connection for two gauges. The high-pressure gauge is included with the manifold; the low-pressure gauge 59620-62 must be ordered separately.
1 Year Warranty
Specifications
  • DescriptionTwo-Gauge Manifold for Manual Hydraulic Press Test