Capp ExpellPlus Sterile, Filtered, Pipette Tips, 100 uL; 10 Racks x 96 Tips Each
Representative image only.
Capp ExpellPlus Sterile, Filtered, Pipette Tips, 100 uL; 10 Racks x 96 Tips Each
Capp | Mfr: 5030066C| Item: 2520194
Accessories and tips for your CAPP® ecopipette™ Single-Channel Variable-Volume Pipettor
₹18,900.00INR / Pkg of 960
₹18,900.00INR / Pkg of 960
Product Description
Key Features
  • “Go Green” and “Save Some Green”
  • Compatible with the 25201-08 and -12 Ecopipette pipettor
More About this Item
These replacement parts and tips are suitable for use with the CAPP® ecopipette™ Single-Channel Variable-Volume Pipettors, models 25201-02 through -16. Pipettors sold separately.
Special polymers and ultra-smooth inner surfaces virtually eliminate sample loss due to retention or binding. Filters minimize the risk of cross-contamination of samples and the pipette. These sterile tips are also certified free of RNase, DNase, pyrogens, RNA, and DNA.
Specifications
  • SterileYes
  • AutoclavableYes
  • Max Volume (µL)100
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)0.1
  • Max Dispensing (mL)0.1
  • Number Of Channels1
  • Color CodeNatural
  • Package Of10 racks; 96 tips per rack
  • Compatible withCAPP® Ecopipette
  • DescriptionSterile, Filtered, Pipette Tips, 100 uL; 10 Racks x 96 Tips Each