Capp ExpellPlus Pipette Tips, 0.2-50 µL, Sterile, No Filter; 384/Rack, 10 Racks/Cs
Representative image only.
Capp ExpellPlus Pipette Tips, 0.2-50 µL, Sterile, No Filter; 384/Rack, 10 Racks/Cs
Capp | Mfr: 5030005C| Item: 2501932
Presterilized tips save time and reduce cross-contamination
₹174,000.00INR / Case of 10
₹174,000.00INR / Case of 10
Buy It Together
Current Item
Capp ExpellPlus Pipette Tips, 0.2-50 µL, Sterile, No Filter; 384/Rack, 10 Racks/Cs
Capp ExpellPlus Pipette Tips, 0.2-50 µL, Sterile, No Filter; 384/Rack, 10 Racks/Cs
2501932
₹174,000.00INR / Case of 10
+
CAPPAero384 MultiChannel Pipettor, 16-Channel, 0.2-2 µL
2501900
₹389,000.00INR / Each
=
Total Price 563000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Compatible with the CAPP® CAPPAero384 multichannel pipettor
  • 3 year warranty
More About this Item
Specially engineered to fit CappAero™ multichannel pipettors. ExpellPlus tips are suitable for CAPP 16, 24, 32, 48 and 64 channel pipettes. These high-accuracy, low-retention tips are designed to minimize sample loss due to stickier substances like DNA, RNA, and proteins.
3 Year Warranty
Specifications
  • SterileYes
  • Max Volume (µL)50
  • Min Volume (µL)0.2
  • Tip Volume (mL)0.05
  • Overall Length (InchShort)1.5
  • Overall Length (mm)38.1
  • Tip StyleLow Retention
  • Color CodeNatural
  • Filter TypeNone
  • Package Of10 racks; 384 tips per rack
  • Compatible withCAPP® CAPPAero384 Mulichannel Pipettors
  • DescriptionExpellPlus Pipette Tips, 0.2-50 µL, Sterile, No Filter; 384/Rack, 10 Racks/Cs