Cannon 9721-F83 Viscometer, universal size 600
Representative image only.
Cannon 9721-F83 Viscometer, universal size 600
Cannon | Mfr: 9721-F83| Item: 9893481
Determine viscosities using ASTM testing methods
₹65,900.00INR / Each
₹65,900.00INR / Each
Buy It Together
Current Item
Cannon 9721-F83 Viscometer, universal size 600
Cannon 9721-F83 Viscometer, universal size 600
9893481
₹65,900.00INR / Each
+ =
Total Price 82100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Each viscometer is calibrated in accordance with ASTM D446 Comes with a calibration certificate that includes the viscometer constant and formulas for calculating viscosities
  • Reverse flow type
  • 1 year warranty
More About this Item
Use for dark Newtonian liquids; especially suited for liquids so dark in color that the meniscus cannot be seen in a Cannon-Fenske routine viscometer. Use to study the effects of various additives on lubricating and hydraulic oils. Also use to study shearing stress and shearing rate. Require a 12-mL sample volume.
1 Year Warranty
Specifications
  • Accuracy±0.3%
  • Viscometer TypeCannon-Fenske Opaque
  • Approximate Constant (cSt/sec)20
  • Sample Volume Needed (mL)12
  • Universal Size Number600
  • Min Viscosity Range (cSt)4000
  • Max Viscosity Range (cSt)20000
  • DescriptionViscometer, universal size 600