Cannon 9721-B80 Viscometer, universal size 500
Representative image only.
Cannon 9721-B80 Viscometer, universal size 500
Cannon | Mfr: 9721-B80| Item: 9893460
Determine viscosities using ASTM testing methods
₹49,400.00INR / Each
₹49,400.00INR / Each
Buy It Together
Current Item
Cannon 9721-B80 Viscometer, universal size 500
Cannon 9721-B80 Viscometer, universal size 500
9893460
₹49,400.00INR / Each
+
Sensidyne 216S Gas Detection Tubes, Acetic acid
8190011
₹23,800.00INR / Box of 10
+ + =
Total Price 98300.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each viscometer is calibrated in accordance with ASTM D446 Comes with a calibration certificate that includes the viscometer constant and formulas for calculating viscosities
  • 1 year warranty
More About this Item
Quickly and easily measure the viscosities of transparent Newtonian liquids in the chemical and petroleum industries. Require a 7-mL sample volume.
1 Year Warranty
Specifications
  • Accuracy±0.2%
  • Viscometer TypeCannon-Fenske Routine
  • Approximate Constant (cSt/sec)8
  • Sample Volume Needed (mL)7
  • Universal Size Number500
  • Min Viscosity Range (cSt)1600
  • Max Viscosity Range (cSt)8000
  • DescriptionViscometer, universal size 500