Caframo Mixer Stand, 12" x 9-1/2" Base with 5/8" dia x 24"L SS Rod

Representative image only.

Caframo Mixer Stand, 12" x 9-1/2" Base with 5/8" dia x 24"L SS Rod

Caframo | Mfr: A210| Item: 5080150

Compact yet sturdy stand for compact mixers

₹65,400.00INR / Each

for an additional
₹39,900.00INR / Each
for an additional
₹37,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹65,400.00INR / Each

Buy It Together

Current Item

Caframo Mixer Stand, 12" x 9-1/2" Base with 5/8" dia x 24"L SS Rod
Caframo Mixer Stand, 12" x 9-1/2" Base with 5/8" dia x 24"L SS Rod
5080150

₹65,400.00INR / Each
+
InnoCal NIST-Traceable Calibration; Mixer
1705003

₹37,400.00INR / Each
=
Total Price 102800.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Cast zinc-aluminum alloy base with coating for corrosion and chemical resistance
  • 2 year warranty

More About this Item

A slight offset design accommodates Caframo clamp so impeller is placed in the center of vessel.

Two year warranty
Specifications
  • MaterialCast zinc-aluminum alloy base with 304 SS rod
  • DescriptionMixer Stand, 12" x 9-1/2" Base with 5/8" dia x 24"L SS Rod
  • Warranty2 years
Reviews
Q&A