Burrell 075-777-00-00 Finger SUREGRIP™ Clamp
Representative image only.
Burrell 075-777-00-00 Finger SUREGRIP™ Clamp
Burrell | Mfr: 075-777-00-00| Item: 6115162
Enhance your wrist-action shakers with accessories
₹30,600.00INR / Each
₹30,600.00INR / Each
Product Description
Key Features
  • Vinyl-sleeved grippers hold almost any size or shape flask or bottle, up to 250 mL
  • Adjustable positioning helps accommodate various sizes and shapes
More About this Item
Clamps accommodate various size flasks and bottles to suit your applications.
Use this versatile clamp to hold flasks, bottles, or containers, up to 250 mL. Easily adjust along the shaft to accommodate a variety of size and shape vessels.
Specifications
  • Compatible WithFlasks, bottles, or containers, up to 250 mL
  • DescriptionFinger SUREGRIP™ Clamp