Burkle 9630-0090 Pipette stand with two rotating plates and drip pan, PP, holds up to 40 pipettes
Representative image only.
Burkle 9630-0090 Pipette stand with two rotating plates and drip pan, PP, holds up to 40 pipettes
Burkle | Mfr: 9630-0090| Item: 0604443
Dry or store pipettes safely, securely, and vertically
₹39,400.00INR / Each
₹39,400.00INR / Each
Product Description
Includes
stand base with 17-3/4" (45 cm) H rod, two perforated clamps, and one drip-pan clamp.
Key Features
  • Compatible with 10 to 18 mm diameter pipettes and thermometers
  • Drip pan protects tips and collects runoff
  • Magnetic clamps can be adjusted with one hand
  • High-capacity to save space—forty slots
  • Chrome-plated steel and polypropylene construction resists corrosion
  • 15 month warranty
More About this Item
Adjust the perforated tray or drip pan height with one hand—magnets have replaced screw-style clamps. Adjust the height of the trays up to 17-3/4" (45 cm). Store and dry up to forty pipettes or thermometers at once—compatible with diameters of 10 to 18 mm.
15 Month Warranty
Specifications
  • Clamp MaterialPolypropylene
  • Rod MaterialChrome-plated steel
  • Base MaterialPolypropylene
  • Clamp TypePipette/Thermometer
  • ColorWhite
  • DescriptionPipette Stand with Clamps, 40-Place; 10-18 mm Diameter
  • Warranty15 month