Bullard EVA/EVAHL Decontamination Kit; 1/Ea
Representative image only.
Bullard EVA/EVAHL Decontamination Kit; 1/Ea
Bullard | Mfr: EVADSK| Item: 1639231
Deep clean your EVA/EVAHL respirators with water submersion
₹7,500.00INR / Each
₹7,500.00INR / Each
Product Description
Includes
PA3PG Breathing Tube Outlet Plug and Gasket 1 nos , EVAICP67 Filter Inlet Plug and gasket 1 nos , white pad and 2 gaskets
Key Features
  • Submerge in standing or running water using Decontamination Kit – clean in under 20 seconds
  • Kit should be replaced every 50 uses
  • Compatible with EVA and EVAHL Respirators
  • Tested with below disinfectant range:
  • o Process NPD (1.256) from Steris
  • o Spor-Klenz
  • o Clorox liquid bleach 10% concentration
  • o 100% Methanol
  • o 70% IPA
Specifications
  • DescriptionEVA/EVAHL Decontamination Kit; 1/Ea