Brookfield AMETEK SSA31/13R Small sample adapter for models 98945-00 and -05; 30 to 100 K cp
Representative image only.
Brookfield AMETEK SSA31/13R Small sample adapter for models 98945-00 and -05; 30 to 100 K cp
Brookfield AMETEK | Mfr: SSA31/13R| Item: 9894558
₹461,000.00INR / Each
₹461,000.00INR / Each
Buy It Together
Current Item
Brookfield AMETEK SSA31/13R Small sample adapter for models 98945-00 and -05; 30 to 100 K cp
Brookfield AMETEK SSA31/13R Small sample adapter for models 98945-00 and -05; 30 to 100 K cp
9894558
₹461,000.00INR / Each
+ =
Total Price 657000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
one sample chamber, one spindle, water jacket, attachment hardware, and storage case.
Key Features
  • 1 year warranty
More About this Item
Use the small sample adapters when measuring low fluid volumes of 8 to 13 mL. Adapters include a water jacket for attachment to an external circulating bath (not included). Easily install or remove sample chamber without disturbing operation of viscometer or circulating bath. Adapters operate from 5 to 212°F (-15 to 100°100).
1 Year Warranty
Specifications
  • Min Viscosity Range (cP)30
  • Max Viscosity Range (cP)100000
  • Max Temperature (° F)212
  • Min Temperature (° F)5
  • Max Temperature (° C)100
  • Min Temperature (° C)-15
  • Sample Volume Needed (mL)8
  • DescriptionSmall sample adapter for models 98945-00 and -05; 30 to 100 K cp