Brookfield AMETEK SC4-31 Spindles for temp control unit and small sample adapters; 30 to 100 K
Representative image only.
Brookfield AMETEK SC4-31 Spindles for temp control unit and small sample adapters; 30 to 100 K
Brookfield AMETEK | Mfr: SC4-31| Item: 9894582
Enhance the versatility of your Brookfield rotational viscometers with these accessories
₹208,000.00INR / Each
₹208,000.00INR / Each
Buy It Together
Current Item
Brookfield AMETEK SC4-31 Spindles for temp control unit and small sample adapters; 30 to 100 K
Brookfield AMETEK SC4-31 Spindles for temp control unit and small sample adapters; 30 to 100 K
9894582
₹208,000.00INR / Each
+ =
Total Price 860000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Select spindles according to your viscometer model and desired viscosity range.
1 Year Warranty
Specifications
  • Min Viscosity Range (cP)30
  • Max Viscosity Range (cP)100000
  • DescriptionSpindles for temp control unit and small sample adapters; 30 to 100 K