Brookfield AMETEK SC4-28 Spindle for temp control unit and small sample adapters; 500 to 1.7 M
Representative image only.
Brookfield AMETEK SC4-28 Spindle for temp control unit and small sample adapters; 500 to 1.7 M
Brookfield AMETEK | Mfr: SC4-28| Item: 9894586
Enhance the versatility of your Brookfield rotational viscometers with these accessories
₹148,000.00INR / Each
₹148,000.00INR / Each
Buy It Together
Current Item
Brookfield AMETEK SC4-28 Spindle for temp control unit and small sample adapters; 500 to 1.7 M
Brookfield AMETEK SC4-28 Spindle for temp control unit and small sample adapters; 500 to 1.7 M
9894586
₹148,000.00INR / Each
+ =
Total Price 800000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Select spindles according to your viscometer model and desired viscosity range.
1 Year Warranty
Specifications
  • Min Viscosity Range (cP)500
  • Max Viscosity Range (cP)1700000
  • DescriptionSpindle for temp control unit and small sample adapters; 500 to 1.7 M