Brookfield AMETEK RVHV-5 Replacement Viscometer Spindle
Representative image only.
Brookfield AMETEK RVHV-5 Replacement Viscometer Spindle
Brookfield AMETEK | Mfr: RVHV-5| Item: 9894578
₹84,500.00INR / Each
₹84,500.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Order replacement spindles according to your viscometer model and desired centipoise range.
1 Year Warranty
Specifications
  • Min Viscosity Range (cP)400
  • Max Viscosity Range (cP)6400000
  • DescriptionReplacement Viscometer Spindle