BriskHeat INSUL2 INSUL-LOCK DS Flexible Pipe Insulation, Closed Cell, 1.5/1.75" dia. pipe, 6-ft L
Representative image only.
BriskHeat INSUL2 INSUL-LOCK DS Flexible Pipe Insulation, Closed Cell, 1.5/1.75" dia. pipe, 6-ft L
BriskHeat | Mfr: INSUL2| Item: 3611553
Cost effective, flexible thermal insulation for pipes
₹8,700.00INR / Each
₹8,700.00INR / Each
Product Description
Key Features
  • Suitable for indoor or outdoor use
  • Easy-to-install
  • 1 year warranty
More About this Item
Suitable for use with SpeedTrace extreme heating cables.
1 Year Warranty
Specifications
  • Max Temperature (° F)220
  • Max Temperature (° C)104
  • Min Temperature-70;-57
  • Plug TypeNo plug
  • Length (ft)6
  • Diameter (in)2
  • Thickness (in)1/2
  • DescriptionINSUL-LOCK DS Flexible Pipe Insulation, Closed Cell, 1.5/1.75" dia. pipe, 6-ft L