BriskHeat CPHTC2030 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 3', 53 Watts, 240 V

Representative image only.

BriskHeat CPHTC2030 Flexible, Small-Diameter, Fiberglass Insulated Heating Cord, 3', 53 Watts, 240 V

BriskHeat | Mfr: CPHTC2030| Item: 0312231

Spiral wound heating element-packs more heating element into each inch of cord.

₹27,200.00INR / Each

₹27,200.00INR / Each
Product Description

Key Features

  • Heat to 850ºF (454ºC)
  • Only 3/16" diameter-ideal for heating in tight spaces
  • 1 year warranty

More About this Item

240 VAC models have stripped ends.

Use this table to determine the best coil spacing for you application. For example, to achieve a maximum temperature of 602 ºF (317ºC) on a 1" OD tubing at 120 VAC, you need to space the heating coils 1/4" apart.

Rugged, small-diameter cords heat small vessels or tubes where space is too limited for heating tapes. Heatglass, ceramic, or metal. Cords are suitable for laboratory, production, or maintenance applications. A tough, woven fiberglass outer sheath protects you from electrical hazards. NiChrome wire, spiral-wound around the core, packs more heating element into the cord.

Control heating cord temperature with our DIGI-SENSE® temperature controller model 89800-02 (sold below right) or vary the output of 240 VAC models with variable-voltage transformers 01575-05 or -15). Heating cords are supplied with two cord ties and 10" leads.

1 Year Warranty
Specifications
  • Plug TypeSplit plug
  • Power (VAC)240
  • Power (watts)53
  • Width (in)3/16
  • Length (ft)3
  • DescriptionFlexible, Small-Diameter, Fiberglass Insulated Heating Cord, 3', 53 Watts, 240 V
  • Warranty1 year
Reviews
Q&A