BrandTech Transferpettor 702852 Positive Displacement Pipettor Tips, PP, 100-500 µL, 10/PK
Representative image only.
BrandTech Transferpettor 702852 Positive Displacement Pipettor Tips, PP, 100-500 µL, 10/PK
BrandTech | Mfr: 702852| Item: 2455075
Color coded to match appropriate sized pipettes
₹15,100.00INR / Pkg of 10
₹15,100.00INR / Pkg of 10
Buy It Together
Current Item
BrandTech Transferpettor 702852 Positive Displacement Pipettor Tips, PP, 100-500 µL, 10/PK
BrandTech Transferpettor 702852 Positive Displacement Pipettor Tips, PP, 100-500 µL, 10/PK
2455075
₹15,100.00INR / Pkg of 10
+ =
Total Price 210100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Robust polypropylene (PP) construction
  • Reusable
More About this Item
Reusable tips are specially designed for the Transferpettor positive displacement pipettor.
Specifications
  • SterileNo
  • Max Volume (µL)500
  • Min Volume (µL)100
  • Min Volume (mL)0.1
  • Max Volume (mL)0.5
  • Tip Volume (mL)0.5
  • Min Dispensing (mL)0.1
  • Max Dispensing (mL)0.5
  • Body MaterialPolypropylene (PP)
  • Color CodeGreen
  • DescriptionPositive Displacement Pipettor Tips, PP, 100-500 uL, 10/pk