BrandTech HandyStep® Touch S Repeating Pipette Bundle
Representative image only.
BrandTech HandyStep® Touch S Repeating Pipette Bundle
BrandTech | Mfr: 705211| Item: 2480603
Next generation, touch screen repetitive pipette with advanced features
₹443,000.00INR / Each
₹443,000.00INR / Each
Buy It Together
Current Item
BrandTech HandyStep® Touch S Repeating Pipette Bundle
BrandTech HandyStep® Touch S Repeating Pipette Bundle
2480603
₹443,000.00INR / Each
+ =
Total Price 468300.00 ADD 2 ITEMS TO CART
Product Description
Includes
Repetitive pipette, charging stand, shelf/rack mount, samplePD-Tip, lithium-ion battery, and universal AC adapter.
Key Features
  • Easy operation with context-sensitive help screens
  • 1 year warranty
More About this Item
The HandyStep® touch pipette is an electronic repeating pipettor with touch-screen operation, ability to store favorites, and motorized tip ejection to fit the needs of your application. It features an intuitive menu navigation, integrated help functions, and constantly displays the most important functions and parameters. The unique auto-dispense operating mode adapts to your dispensing rhythm without the need for programming. Motorized tip ejection by the press of a button prevents contamination and upholds sample integrity. Additional features found in the "S" model include sequential dispensing, multi-aspiration, and titration. The automatic tip size recognition of PD-Tip™ II pipette tips saves you time and prevents errors – simply insert the tip and the size is automatically recognized and displayed. When a new PD- Tip™ II tip of the same size is inserted, all settings are maintained. The USB-C connection allows for charging by cable; or inductively charge with the charging stand included in bundles.
1 Year WarrantyConformite Europeenne
Specifications
  • AutoclavableNo
  • Min Volume (µL)1
  • Max Volume (mL)50
  • Number Of Channels1
  • Tip EjectorAutomatic
  • Body MaterialPC/PBT, PP, silicone, glass, PEEK
  • Auto ShutoffYes, after 10 minutes
  • Battery1 3.7V Lithium-ion (included)
  • DescriptionHandyStep® Touch S Repeating Pipette Bundle