BrandTech HandyStep® S Repeating Pipette
Representative image only.
BrandTech HandyStep® S Repeating Pipette
BrandTech | Mfr: 705110| Item: 2480605
Positive displacement principle provides a high degree of accuracy and reproducibility
₹123,000.00INR / Each
₹123,000.00INR / Each
Product Description
Key Features
  • Integrated volume table for quick reference
  • Spring-loaded tip ejection avoids hand contact with contaminated tips
  • Compatible with standard repeating tips
  • 1 year warranty
More About this Item
The HandyStep® S is a manual repeating pipette offering a slim, lightweight design and convenient one-handed operation. In conjuction with the ten sizes of PD- Tip™ tips, it provides 59 different volume settings. The tip ejection system allows you to easily remove tips without handling to prevent contamination. A convenient integrated volume table provides a quick reference while dispensing.
1 Year Warranty
Specifications
  • Number Of Channels1
  • Tip EjectorManual ejection key
  • BatteryNone
  • DescriptionHandyStep® S Repeating Pipette