BrandTech Discharge Tube for Seripettor® Bottletop Dispenser, Volume 10 and 25mL
Representative image only.
BrandTech Discharge Tube for Seripettor® Bottletop Dispenser, Volume 10 and 25mL
BrandTech | Mfr: 704520| Item: 0791568
Renew your dispenser with these original equipment parts and accessories!
₹13,400.00INR / Each
₹13,400.00INR / Each
Product Description
Includes
closure cap, EPDM discharge valve and sealing ring.
Key Features
  • Compatible with the BrandTech® Seripettor® and Serripettor® Pro bottletop dispensers
  • Suitable for use with the Seripettor® bottletop dispenser
More About this Item
Keep your dispenser working at its best with these original BRAND® parts and accessories. Note, some parts are specific to the seripettor® or seripettor® pro, and also the volume range.
Specifications
  • Capacity (mL)10 and 25
  • MaterialPolypropylene and EPDM
  • Compatible WithSeripettor® Bottletop Dispenser
  • DescriptionDischarge Tube for Seripettor® Bottletop Dispenser, Standard Tip, 10 and 25 mL