BrandTech BRANDplates® pureGrade™ Non-Treated, Non-Sterile, Divisible Strip Plate, Clear with Grid, 96-Well, 360 µL, Standard F-Bottom; 100/PK
Representative image only.
BrandTech BRANDplates® pureGrade™ Non-Treated, Non-Sterile, Divisible Strip Plate, Clear with Grid, 96-Well, 360 µL, Standard F-Bottom; 100/PK
BrandTech | Mfr: 782301| Item: 0790427
Medium binding plates for a variety of applications
₹268,000.00INR / Pkg of 100
₹268,000.00INR / Pkg of 100
Buy It Together
Current Item
BrandTech BRANDplates® pureGrade™  Non-Treated, Non-Sterile, Divisible Strip Plate, Clear with Grid, 96-Well, 360 µL, Standard F-Bottom; 100/PK
BrandTech BRANDplates® pureGrade™ Non-Treated, Non-Sterile, Divisible Strip Plate, Clear with Grid, 96-Well, 360 µL, Standard F-Bottom; 100/PK
0790427
₹268,000.00INR / Pkg of 100
+ =
Total Price 319700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Available nonsterile or sterile for specific applications
  • White (luminescence) black (fluorescence) plates available either solid, or with clear bottom for top or bottom reading
  • Choose from three distinct bottom shapes: round, flat, or C (flat, with curved edge)
  • Offered in 96- and 384-well formats
  • Meets ANSI/SLAS dimensions and molded to exacting standards
  • pureGrade™ Divisible Strip Plate, 96-Well Clear without Grid Non-treated and Non-Sterile Volume: 360 µL Standard F-Bottom
More About this Item
These sterile and nonsterile, medium binding, non-treated surface microplates are ideal for covering many standard applications, such as homogenous assays, screening, and storage. The polystyrene microplates are designed to lessen cross contamination, ensure outstanding recovery, and offer many options with color and number of wells. Three different bottom shapes are available: round (U) with no edges, flat (F) with a flat bottom, and flat with a curved edge (C) bottom for enhanced mixing.
With a chimney well type of design, each well is separated from each other to reduce the risk of cross contamination. The 384-well microplates feature rounded well corners which allow optimal uniform meniscus formation.
Strip plates, as well as a handful of convenient accessories for use with the microplates, such as lids, rubber rollers, and sealing films are offered.
Specifications
  • Number of Wells96
  • Well Volume (µL)360
  • Well ShapeStandard F-Bottom
  • Surface TreatmentpureGrade™ Non-treated
  • SterileNon-Sterile
  • Plate ColorClear with grid
  • MaterialPolystyrene
  • DescriptionBrandTech 782301 BRANDplates® pureGrade™ Non-Treated, Non-Sterile, Divisible Strip Plate, Clear with Grid, 96-Well, 360 µL, Standard F-Bottom; 100/PK