BrandTech BIO-CERT® 8-Strip PCR Tubes without Caps, Polypropylene, 0.2 mL, Clear; 125/Pk
Representative image only.
BrandTech BIO-CERT® 8-Strip PCR Tubes without Caps, Polypropylene, 0.2 mL, Clear; 125/Pk
BrandTech | Mfr: 781320| Item: 0790546
High quality polymerase chain reaction plastics free from leachables
₹23,400.00INR / Pkg of 125
₹23,400.00INR / Pkg of 125
Product Description
Key Features
  • BIO-CERT® PCR QUALITY—Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
  • Thin uniform wall thickness for precise thermal transfer
  • Clear 8-strip PCR tubes without caps
More About this Item
These PCR tube strips are made of top-quality virgin resins without lubricants or chemical additives. They are an excellent choice for producing reproducible results in nearly all PCR applications.
Specifications
  • Number of Wells8
  • Well Volume (mL)0.2
  • Lid (Included)No
  • SterileNo
  • Plate Type8-Strip PCR Tube
  • Plate ColorClear
  • MaterialPolypropylene
  • Description8-Strip PCR Tubes without Caps, Polypropylene, 0.2 mL, Clear; 125/Pk