BrandTech 759243 Round Caps for Ultra-Micro UV-Cuvettes, Orange, 100/pack
Representative image only.
BrandTech 759243 Round Caps for Ultra-Micro UV-Cuvettes, Orange, 100/pack
BrandTech | Mfr: 759243| Item: 0634393
Optimize usage of your disposable cuvettes
₹9,700.00INR / Pkg of 100
₹9,700.00INR / Pkg of 100
Product Description
Key Features
  • Virgin raw polyethylene (PE) resists common lab reagents
  • Color-coded options support fast sample identification and workflow organization
  • Flexible round design provides uniform coverage with easy placement and removal
  • Manufactured under controlled conditions with batch-managed quality inspection
More About this Item
These disposable round caps support UV cuvette workflows by helping protect samples during handling and measurement.
Manufactured from virgin raw polyethylene under controlled conditions, each cover provides a clean, consistent fit that helps reduce evaporation and airborne contamination without interfering with optical readings.
The flexible material allows quick placement and removal, making them well suited for life science, analytical, and routine lab applications. Color-coded options further streamline sample identification and organization in busy labs.
Specifications
  • MaterialPolyethylene (PE)
  • ColorOrange
  • Compatible WithBrandTech disposable ultra-micro-volume cuvettes
  • DescriptionRound Caps for Ultra-Micro UV-Cuvettes, Orange, 100/pack