BrandTech 732002 Bulk nano-cap™ Pipette Tips, 0.1-20 uL; 2 Bags, 1000/ea
Representative image only.
BrandTech 732002 Bulk nano-cap™ Pipette Tips, 0.1-20 uL; 2 Bags, 1000/ea
BrandTech | Mfr: 732002| Item: 2455497
Pipette tips fit BRAND® Transferpette® and most major brands
₹29,200.00INR / Pkg of 2000
₹29,200.00INR / Pkg of 2000
Product Description
Key Features
  • Available non-sterile or BIO-CERT® sterile
  • Oleamide and diHEMDA-free
  • Rack packed tips up to 1000µL free of DNA, RNase, endotoxin and ATP to detection limit
  • Sterile tips have a Sterility Assurance Level (SAL) of 10-6
  • Ideal for molecular biological applications, such as PCR
  • 37 mm length features capillary rise that is visible at 0.1 uL
  • Capillary part of tip fills gel wells for most gel electrophoresis systems
  • Suitable for pipettes up to 20 uL
  • Racked tips are colorless in gray tip tray
More About this Item
BRAND® pipette tips are made of a high-purity polypropylene that is free of additives, lubricants and pigments (bulk 200µL and 1000µL tips feature cadminum-free pigments for easy identification) that cause sample contamination. All tips are manufactured in a clean room under state-of-the-art production conditions and automatically packaged to ensure high quality. Tips can be autoclaved at 121°C (250°F) and 15 psi (1 bar). Choose between non-sterile and BIO-CERT® sterile tips, which adhere to ISO 11137 and AAMI guidelines.
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)20
  • Min Volume (µL)0.1
  • Min Volume (mL)0.0001
  • Max Volume (mL)0.02
  • Overall Length (InchShort)1.5
  • Overall Length (mm)37
  • Body MaterialPolypropylene
  • Color CodeColorless in gray tip tray
  • DescriptionBrandTech 732002 BRAND® Bulk Nano-Cap™ Pipette Tips, 0.1-20 uL; 2 Bags, 1000/ea