BrandTech 707930 Drying Tube for Dispensette® and Titrette® Dispensers
Representative image only.
BrandTech 707930 Drying Tube for Dispensette® and Titrette® Dispensers
BrandTech | Mfr: 707930| Item: 0787853
Helps maintain dry, contamination-free dispensing
₹18,300.00INR / Each
for an additional
₹14,500.00INR / Each
for an additional
₹25,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹18,300.00INR / Each
Buy It Together
Current Item
BrandTech 707930 Drying Tube for Dispensette® and Titrette® Dispensers
BrandTech 707930 Drying Tube for Dispensette® and Titrette® Dispensers
0787853
₹18,300.00INR / Each
+ =
Total Price 32800.00 ADD 2 ITEMS TO CART
Product Description
Includes
Valve block sealing ring
Key Features
  • Protect sensitive reagents from humidity and carbon dioxide
More About this Item
An optional drying tube screws into the accessory port of a Dispensette® or Titrette® dispenser to protects sensitive reagents from humidity and carbon dioxide. Fill drying tube with solid reagent that is suitable for the application. Add a bit of mineral wool into the tube as a filter material to prevent the solid reagent or its dust from entering the bottle.
Specifications
  • Compatible WithDispensette® and Titrette® dispensers
  • DescriptionDrying Tube for Dispensette® and Titrette® Dispensers