BrandTech 704399 Threaded Bottle Adapter, ETFE, GL45 Thread x GL38 Bottle Thread
Representative image only.
BrandTech 704399 Threaded Bottle Adapter, ETFE, GL45 Thread x GL38 Bottle Thread
BrandTech | Mfr: 704399| Item: 0790208
Adapt dispenser to bottles with various thread sizes for your application needs
₹11,700.00INR / Each
₹11,700.00INR / Each
Product Description
Key Features
  • Easily transition from your dispenser to your bottle
  • 45/38 mm ETFE threaded adapter to fit many common gallon/4 liter bottles
  • 1 year warranty
More About this Item
45/38 mm ETFE thread adapter to fit many common gallon/4 liteer bottles.
For bottle adapters with a bottle thread diameter of less than 32 mm, the bottle adapter 07910-95 must be inserted.
1 Year Warranty
Specifications
  • MaterialETFE
  • Compatible WithDispensette®, Seripettor®, and Titrette® dispensers
  • ConnectionsGL45 Thread x GL38 Bottle Thread
  • DescriptionThreaded Bottle Adapter, ETFE, GL45 Thread x GL38 Bottle Thread