BrandTech 704325 Threaded Bottle Adapter, PP, GL32 Thread x GL25 Bottle Thread
Representative image only.
BrandTech 704325 Threaded Bottle Adapter, PP, GL32 Thread x GL25 Bottle Thread
BrandTech | Mfr: 704325| Item: 0791078
Adapt dispenser to bottles with various thread sizes for your application needs
₹3,400.00INR / Each
₹3,400.00INR / Each
Product Description
Key Features
  • Easily transition from your dispenser to your bottle
  • 32/25 mm PP threaded adapter to fit many common bottles
More About this Item
For bottle adapters with a bottle thread diameter of less than 32 mm, the bottle adapter 07910-95 must be inserted.
Specifications
  • MaterialPolypropylene
  • Compatible WithDispensette®, Seripettor®, and Titrette® dispensers
  • ConnectionsGL32 Thread x GL25 Bottle Thread
  • DescriptionThreaded Bottle Adapter, PP, GL32 Thread x GL25 Bottle Thread