BrandTech 388050 Rack for 2 1000 mL Imhoff Sedimentation Cones, PMMA; 1/EA
Representative image only.
BrandTech 388050 Rack for 2 1000 mL Imhoff Sedimentation Cones, PMMA; 1/EA
BrandTech | Mfr: 388050| Item: 0610239
Easy drainage with bottom screw on cap
₹120,000.00INR / Each
₹120,000.00INR / Each
Product Description
Key Features
  • 1000 mL volume
  • Graduated per DIN 12672
  • Withstands temperatures up to 85° C
More About this Item
This settling cone is made of styrene acrylonitrile to offer exceptional optical clarity for measuring of settled solids.