BrandTech 23728 Bottle Base for Dr. Schilling (Automatic) Burettes, Fits 1000 mL Bottle with 94 to 96 mm Dia.

Representative image only.

BrandTech 23728 Bottle Base for Dr. Schilling (Automatic) Burettes, Fits 1000 mL Bottle with 94 to 96 mm Dia.

BrandTech | Mfr: 23728| Item: 0791075

Maximize safety, convenience and productivity

₹5,900.00INR / Each

₹5,900.00INR / Each
Product Description

Key Features

  • Fits 1000 mL bottle with 94 to 96 mm diameter

More About this Item

Use the base to help stabilize your automatic burette dispensing bottle.

Specifications
  • MaterialHDPE
  • Compatible WithSchilling automatic burette; 1000 mL plastic bottle with a diameter of 94 to 96 mm
  • Description23728 Bottle Base for Dr. Schilling (Automatic) Burettes, Fits 1000 mL Bottle with 94 to 96 mm Dia.
Reviews
Q&A