BrandTech 23725 Bottle Base for Dr. Schilling (Automatic) Burettes, Fits 500 mL Bottle with 74 to 76 mm Dia

Representative image only.

BrandTech 23725 Bottle Base for Dr. Schilling (Automatic) Burettes, Fits 500 mL Bottle with 74 to 76 mm Dia

BrandTech | Mfr: 23725| Item: 0791073

Maximize safety, convenience and productivity

₹5,900.00INR / Each

₹5,900.00INR / Each
Product Description

Key Features

  • Fits 500 mL bottle with 74 to 76 mm diameter

More About this Item

Use the base to help stabilize your automatic burette dispensing bottle.

Specifications
  • MaterialHDPE
  • Compatible WithSchilling automatic burette; 500 mL plastic bottle with a diameter of 74 to 76 mm
  • Description23725 Bottle Base for Dr. Schilling (Automatic) Burettes, Fits 500 mL Bottle with 74 to 76 mm Dia
Reviews
Q&A