BrandTech 23320 Reservoir Bottle for Titrette® Bottletop Burette, Amber, 2L (Requires Adapter)

Representative image only.

BrandTech 23320 Reservoir Bottle for Titrette® Bottletop Burette, Amber, 2L (Requires Adapter)

BrandTech | Mfr: 23320| Item: 0791071

Maximize convenience and productivity

₹113,000.00INR / Each

₹113,000.00INR / Each
Product Description

Key Features

  • Use with Titrette® burette and bottle adapter to accurately dispense for titration
  • Requires 33 mm-NS 29/32 adapter (supplied with Titrette® burette)
  • Perfect for light-sensitive reagents
Specifications
  • Capacity (Liters)2
  • MaterialBorosilicate glass 3.3
  • ColorAmber
  • Height (cm)20
  • Diameter (cm)16
  • Description23320 Reservoir Bottle for Titrette® Bottletop Burette, Amber, 2L (Requires Adapter); 1/EA
Reviews
Q&A